Learn More
Quidel Corporation MicroVue BAP EIA Assay Kit
Supplier: Quidel Corporation 8012
The MicroVue BAP EIA provides a quantitative measure of bone-specific alkaline phosphatase (BAP) activity in serum.