Learn More
BioworldSupplier Diversity Partner Mid/Low-Range Protein Marker (14.4-97.4kDa), 1 mL DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 220100012
Mid/Low range protein molecular weight markerSpecification:Ultra Pure GradeNumber of Bands: 7 Bands Marker Range:14.4-97.4 KDa
Catalog No. 50-252-979