Learn More
SPEX™ SamplePrep Mid-Size Grinding Vial Set
Ideal for grinding samples such as plant and animal tissue, seeds, polymers, pharmaceuticals, textiles, hair, and bones
$218.00 - $1198.00
Requires: 886 Mid-Size Grinding Vial Adapter for grinding in the 6870 Large Freezer/Mill and 6884 Mid-Size Grinding Vial Adapter for Large Extractor/Vial Opener.
Products 4
Catalog Number Mfr. No. For Use With (Equipment) Product Type Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Product Type Price Quantity & Availability  
68-81
View Documents
SPEX™ SamplePrep
6881
Large Extractor/Vial Opener Conjunction with Mid Size Grinding Vial Adapter Grinding Vial Set
Each for $1,198.00
Add to cart
68-81P
View Documents
SPEX™ SamplePrep
6881P
Mid Size Grinding Vial Set Impactor
Each for $218.00
Add to cart
68-81C-20
View Documents
SPEX™ SamplePrep
6881C20
Use with 6881 Vial Set Cylinder
Case of 20 for $1,087.00
Add to cart
68-83
View Documents
SPEX™ SamplePrep
6883
Large Freezer/Mill (Mfr. No. 6875) and Large Dual Freezer/Mill (Mfr. No. 6875 D), When Used with Mid-Size Grinding Vial Adapter (6887) Mid-Size Cr-free Vial
Each for $970.00
Add to cart
Description
Description
Mid-Size Grinding Vial Set contains everything necessary to make one complete grinding vial plus three spare polycarbonate enter cylinders.
  • Grinds one sample at a time in the Large Freezer/Mill™ when used in conjunction with the 6886 Mid-Size Grinding Vial Adapter
  • Sample grinding capacity: 3 to 25 g (25 mL)
  • Withstands liquid nitrogen temperatures
  • Closed grinding vial prevents sample cross-contamination, retains volatiles, and controls hazardous or critical samples
  • Transparent plastic center cylinder allows visual checking of the grinding progress
  • Stainless-steel end plugs and impactor are autoclavable and reusable
  • Polycarbonate center cylinders are disposable or may be cleaned with hot water and detergent or RNase/DNase-free solution
  • Compatible with 6804 Large Extractor/Vial Opener when used in conjunction with 6884 Mid-Size Grinding Vial Adapter