Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
midA Mouse anti-Human, Mouse, DyLight 350, Clone: OTI4A5, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272681UV
Description
midA Monoclonal specifically detects midA in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
midA | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
NDUFAF7 | |
Full length recombinant protein of human midA (NP_653337) produced in HEK293T cells. | |
0.1 mL | |
55471.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI4A5 | |
Western Blot, Immunohistochemistry | |
chromosome 2 open reading frame 56, MidA, mitochondrial, PRO1853 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse | |
IgG2b |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title