Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Midecamycin, 900μg/mg

Click to view available options
Quantity:
500mg
1g
35457-80-8
C41H67NO15
813.979
MFCD00865000
DMUAPQTXSSNEDD-RBATUFKESA-N
124225192
[(4R,5R,6S,7R,9R,10R,11E,13Z,16R)-6-[(2S,3R,4R,5S,6R)-4-(dimethylamino)-3-hydroxy-5-[(2S,4R,5S,6S)-4-hydroxy-4,6-dimethyl-5-propanoyloxyoxan-2-yl]oxy-6-methyloxan-2-yl]oxy-10-hydroxy-5-methoxy-9,16-dimethyl-2-oxo-7-(2-oxoethyl)-1-oxacyclohexadeca-11,13-di
CCC(=O)OC1CC(=O)OC(CC=CC=CC(C(CC(C(C1OC)OC2C(C(C(C(O2)C)OC3CC(C(C(O3)C)OC(=O)CC)(C)O)N(C)C)O)CC=O)C)O)C
35457-80-8
C41H67NO15
813.979
MFCD00865000
DMUAPQTXSSNEDD-RBATUFKESA-N
124225192
[(4R,5R,6S,7R,9R,10R,11E,13Z,16R)-6-[(2S,3R,4R,5S,6R)-4-(dimethylamino)-3-hydroxy-5-[(2S,4R,5S,6S)-4-hydroxy-4,6-dimethyl-5-propanoyloxyoxan-2-yl]oxy-6-methyloxan-2-yl]oxy-10-hydroxy-5-methoxy-9,16-dimethyl-2-oxo-7-(2-oxoethyl)-1-oxacyclohexadeca-11,13-di
CCC(=O)OC1CC(=O)OC(CC=CC=CC(C(CC(C(C1OC)OC2C(C(C(C(O2)C)OC3CC(C(C(O3)C)OC(=O)CC)(C)O)N(C)C)O)CC=O)C)O)C

A broad spectrum antibiotic

Solubility Information Soluble in methanol or ethanol; Insoluble in water
Formula Weight 813.97
Physical Form Powder
Boiling Point 874°C
Color White
Melting Point 155°C to 156°C
Quantity 1g
Chemical Name or Material Midecamycin, 900μg/mg

RTECSNumber : OH4730600

storageNote1 : 2° to 8°C