Learn More
Midodrine hydrochloride, MilliporeSigma™ Supelco™
Midodrine is a prodrug and a long acting α-adrenergic stimulating agent, and forms an active metabolite, desglymidodrine, which causes an elevation of the systemic blood pressure with the reduction in heart rate.
Supplier: MilliporeSigma Supelco M82771G
Description
Featured Industries: Pharmaceutical (small molecule)Specifications
3092-17-9 | |
Analytical Standard | |
MFCD00079455 | |
290.74 | |
Drug Analysis |
1 g | |
C12H18N2O4 · HCl | |
(+/-)-1-(2,5-Dimethoxyphenyl)-2-glycinamidoethanol | |
Analytical Standard | |
Neat |