Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Midodrine hydrochloride, MilliporeSigma™ Supelco™
Midodrine is a prodrug and a long acting α-adrenergic stimulating agent, and forms an active metabolite, desglymidodrine, which causes an elevation of the systemic blood pressure with the reduction in heart rate.
$139.00 - $445.00
Specifications
CAS | 3092-17-9 |
---|---|
Type | Analytical Standard |
Linear Formula | C12H18N2O4 · HCl |
MDL Number | MFCD00079455 |
Synonym | (+/-)-1-(2,5-Dimethoxyphenyl)-2-glycinamidoethanol |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
11-101-9176
|
MilliporeSigma Supelco
M827750MG |
50 mg |
Each for $139.00
|
|
|||||
11-101-8235
|
MilliporeSigma Supelco
M82771G |
1 g |
Each for $445.00
|
|
|||||
Description
Featured Industries: Pharmaceutical (small molecule)Specifications
3092-17-9 | |
C12H18N2O4 · HCl | |
(+/-)-1-(2,5-Dimethoxyphenyl)-2-glycinamidoethanol | |
Analytical Standard | |
Neat |
Analytical Standard | |
MFCD00079455 | |
290.74 | |
Drug Analysis |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title