Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MIER2 Antibody (OTI3G7), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272683UV
Description
MIER2 Monoclonal specifically detects MIER2 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| MIER2 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MIER2 | |
| Recombinant protein fragment corresponding to amino acids 1-296 of human MIER2 (NP_060020) produced in E.coli. | |
| 0.1 mL | |
| 54531.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI3G7 | |
| Western Blot, Immunohistochemistry | |
| KIAA1193mesoderm induction early response protein 2, mesoderm induction early response 1, family member 2, Mi-er2 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction