Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MIER2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550451

Catalog No. PA550451


Add to cart

Description

Description

The antibody detects endogenous levels of total MIER2 protein.

Transcriptional repressor.
Specifications

Specifications

MIER2
Polyclonal
Unconjugated
MIER2
Liquid
MIER2
Fusion protein corresponding to a region derived from internal residues of human mesoderm induction early response 1, family member 2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
2.2 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
Q8N344
KIAA1193
Rabbit
IgG
100 μL
-20°C
Primary
54531
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit