Learn More
Alfa Aesar™ MIF Antagonist, ISO-1
A cell-permeable isoxazoline compound
Manufacturer:  Alfa Aesar™ J64947MB
 View more versions of this product
Catalog No. AAJ64947MB
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
25mg
−30° to −10°C
A cell-permeable isoxazoline compound
C12H13NO4
(S,R)-3-(4-Hydroxyphenyl)-4,5-dihydro-5-isoxazole acetic acid, methyl ester; Macrophage Migration Inhibitory Factor Antagonist
Signal Transduction Reagent
Pale Yellow
Solid
478336-92-4
235.24g/mol
Light Sensitive