Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MIF Antagonist, ISO-1

A cell-permeable isoxazoline compound

$99.10 - $392.00

Specifications

CAS 478336-92-4
Formula Weight 235.24g/mol
Synonym (S,R)-3-(4-Hydroxyphenyl)-4,5-dihydro-5-isoxazole acetic acid, methyl ester; Macrophage Migration Inhibitory Factor Antagonist
Storage Requirements −30° to −10°C
Format Solid
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ64947LB0 View Documents Alfa Aesar™
J64947LB0
5mg Each for $99.10
Order in Bulk

Request bulk or custom formats

Add to cart
 
AAJ64947MB View Documents Alfa Aesar™
J64947MB
25mg Each for $392.00
Order in Bulk

Request bulk or custom formats

Add to cart
 
Specifications

Specifications

478336-92-4
(S,R)-3-(4-Hydroxyphenyl)-4,5-dihydro-5-isoxazole acetic acid, methyl ester; Macrophage Migration Inhibitory Factor Antagonist
Solid
A cell-permeable isoxazoline compound
Signal Transduction Reagent
235.24g/mol
−30° to −10°C
Light sensitive
Pale Yellow
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C; Store under Argon

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit