Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MIF4GD Mouse anti-Human, Clone: OTI4C11, liquid, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene TA504443
Description
MIF4GD is a protein which contains an MIF4G domain.This gene encodes a protein which contains an MIF4G domain.Specifications
MIF4GD | |
Monoclonal | |
0.78 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
A9UHW6 | |
AD023, MIFD, SLIP1 | |
Mouse | |
IgG1 | |
100 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
57409 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI4C11 | |
Unconjugated | |
MIF4GD | |
Liquid | |
MIF4GD | |
Full length human recombit protein of human MIF4GD produced in HEK293T cell. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title