Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MIG7 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550707

Catalog No. PA550707


Add to cart

Description

Description

The antibody detects endogenous levels of total MIG7 protein.

MIG7 is a cysteine-rich protein found in cell membranes and cytoplasm of carcinoma cells. Tumor tissue and blood from over 200 cancer patients have been shown to express MIG7, regardless of tissue origin. In contrast, it has not been detectable in tissues or blood from normal subjects. Expression of MIG7 seems to be restricted to carcinoma cells and putatively to the early placenta (Cytotrophoblast) and it ft.s expression is important for the invasive capabilities of tumor cells.
Specifications

Specifications

MIG7
Polyclonal
Unconjugated
MIG7
mig-7, MIG7
Rabbit
IgG
100 μL
-20°C
Primary
723788
Immunohistochemistry (Paraffin)
2.4 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
Liquid
MIG7
Synthetic peptide corresponding to a region derived from internal residues of human mig-7
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit