Learn More
GE Healthcare Mighty Small Multiple Gel Caster Available on GSA/VA Contract for Federal Government customers only.
For use with Mini-Vertical Unit SE 250.
Manufacturer:  GE Healthcare 80614251
Catalog No. 45-002-434
Add to cart
Description
Description
  • Casts up to 10 gels at a time
  • Can be used to cast 10 x 8cm glass plates
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Cast up to 10 gels
Complete packsize
Mini-Vertical Unit SE 250
20 x Glass plate, rectangular (10x8 cm), 10 x Aluminium plate, notched (10x8 cm), 2 x Clamps, 100 x Wax paper sheet, 1 x Space saver plate, 5 x Filler sheet, 1 x Filler plug set, 1 x Space-mate assembly template, supplied with the Gel caster
0.75 mm, 1.0 mm and 1.5 mm
Mighty Small Multiple Gel Caster, for SE 250