Learn More
Tocris Bioscience™ Miglitol Available on GSA/VA Contract for Federal Government customers only.
α-glucosidase inhibitor
Manufacturer:  Tocris Bioscience™ 4985/50
Catalog No. 49-855-0R
Add to cart
This item is not returnable. View return policy
Description
Description
Inhibitor of α-glucosidase; antihyperglycemic. Suppresses postprandial hyperglycemia in vivo and reduces plasma glucose concentration in normal rats and in several animal models of diabetes. Shown to inhibit the loss of pancreatic β cells in type 2 diabetic rats. Also inhibits apoptosis and mitochondrial overproduction of reactive oxygen species in endothelial cells in vitro.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Miglitol
>98
50mg
Miglitol
207.22