Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Milameline hydrochloride, Tocris Bioscience™

Catalog Number p-4784148
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
139886-04-7
C8H15ClN2O
190.671
WEBMRZODPLSRKR-MLBSPLJJSA-N
milameline hydrochloride, unii-w867e71lpd, milameline hydrochloride usan, milameline, miramelin, 1,2,5,6-tetrahydro-1-methylnicotinaldehyde e-o-methyloxime monohydrochloride, c8h14n2o.hcl, 3-pyridinecarboxaldehyde, 1,2,5,6-tetrahydro-1-methyl-, o-methyloxime, monohydrochloride, e
9571001
(E)-N-methoxy-1-(1-methyl-3,6-dihydro-2H-pyridin-5-yl)methanimine;hydrochloride
CN1CCC=C(C1)C=NOC.Cl
This item is not returnable. View return policy
139886-04-7
C8H15ClN2O
190.671
WEBMRZODPLSRKR-MLBSPLJJSA-N
milameline hydrochloride, unii-w867e71lpd, milameline hydrochloride usan, milameline, miramelin, 1,2,5,6-tetrahydro-1-methylnicotinaldehyde e-o-methyloxime monohydrochloride, c8h14n2o.hcl, 3-pyridinecarboxaldehyde, 1,2,5,6-tetrahydro-1-methyl-, o-methyloxime, monohydrochloride, e
9571001
(E)-N-methoxy-1-(1-methyl-3,6-dihydro-2H-pyridin-5-yl)methanimine;hydrochloride
CN1CCC=C(C1)C=NOC.Cl

Muscarinic receptor agonist

Muscarinic receptor agonist that displays roughly equal affinity at all receptor subtypes (Ki values are 2.3, 2.4, 3.6, 3.8 and 4.3 μM for hM1, hM2, hM3, hM4 and hM5 receptors respectively). Cognitive enhancer; reverses spatial memory deficits in rats.
Quantity 10 mg
Formula Weight 190.67
Percent Purity >99%
Chemical Name or Material Milameline hydrochloride
Recommended Storage Desiccate at -20°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit