Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Milestone Medical FineFix Formalin-Free Fixative

The molecular fixative of choice for optimum H & E, HC, IHC plus full recovery of nucleic acids

Manufacturer:  Milestone Medical 70147C

 View more versions of this product

Catalog No. 22-310-294


Add to cart

Description

Description

  • For use with Milestone Pathos Delta and other Milestone Medical tissue processors
  • Removes formalin from your lab
  • Low toxicity

Specifications

Specifications

Storage and transport of specimens in cold climate
FineFIX Tissue Processing Solution
4 x 1 gal.
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit