Learn More
Milestone Medical ProWAVE Dehydrating/Clearing Solution
For use with low-fat specimens
Manufacturer:  Milestone Medical 51411C
 View more versions of this product
Catalog No. 22-310-289
Add to cart
Description
Description
  • Available with or without status color indicator
  • Simple one-step dehydration and clearing
  • Eliminates exposure to and disposal costs of xylene
Specifications
Specifications
Low-fat specimens
without Color indicator
ProWAVE™ Solution
4 x 1 gal.