Milestone Medical ProWAVE Dehydrating/Clearing Solution
For use with low-fat specimens
Manufacturer: Milestone Medical TechnologiesSupplier Diversity Partner 51410G
 View all options for this product
Catalog No. 22-310-285
  Check Availability
Add to Cart
    Includes with Color indicator
    For Use With (Application) Low-fat specimens
    Product Type ProWAVE™ Solution
    Quantity 1 gal.
    • Available with or without status color indicator
    • Simple one-step dehydration and clearing
    • Eliminates exposure to and disposal costs of xylene
  • Description & Specifications
Related Products
Product Content Feedback