Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Milk Fat Globule 1, Human (HMFG1) Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with the Thermo Scientific™ Milk Fat Globule 1, Human (HMFG1) Ab-1, Mouse Monoclonal Antibody.

Specifications

Antigen Milk Fat Globule 1, Human (HMFG1) Ab-1
Clone EDM45
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Host Species Murine
View More Specs

Products 3
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS512P
View Documents
Thermo Scientific™
MS512P
Purified Ab with BSA and Azide 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS512R1
View Documents
Thermo Scientific™
MS512R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS512P1
View Documents
Thermo Scientific™
MS512P1
Purified Ab with BSA and Azide 500μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with the Thermo Scientific™ Milk Fat Globule 1, Human (HMFG1) Ab-1, Mouse Monoclonal Antibody.

Human milk fat globule membrane protein (HMFG1) is present on normal human breast epithelial cells and cell lines derived from breast carcinomas, as well as on the outer surface of the human milk fat globule. HMFG1 is considered as a differentiation marker. It is useful as specific breast epithelial marker in normal and neoplastic mammary development.

Host Species: Mouse

Clone: EDM45

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with rat. Others not known.

Epitope: Not determined

Immunogen: Delipidated human milk fat globule membrane preparation

Molecular Weight: 40-45kDa

Positive Control: SKBR-3 cells; Breast, colon, or lung tissues

Cellular Localization: Membrane and cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Milk Fat Globule 1, Human (HMFG1) Ab-1
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Murine
Cancer and Tumor Biology
Delipidated human milk fat globule membrane preparation
EDM45
Monoclonal
RUO
Primary
IgG2a kappa
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit