Learn More
Eisco™ Milky Quartz Mineral Specimen Products for Science Education
Milky Quartz Mineral Specimen has a tightly packed framework and hardness of 7, complete lack of cleavage planes, and distinct euhedral crystal habit.
$2.30 - $28.00
Specifications
Specimen Type Milky Quartz
Type Mineral
View More Specs
Products 3
Catalog Number Mfr. No. Specimen Size Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Specimen Size Quantity Price Quantity & Availability  
S24710 Products for Science Education View Documents Eisco™
ESNG0025
2 to 3 cm 1 Ea. Each for $2.30
Add to cart
S26608 Products for Science Education View Documents Eisco™
ESNG0025PK10
2 to 3 cm 10/Pk. Each for $5.00
Add to cart
S24711 Products for Science Education View Documents Eisco™
ESNG0025PK12
2 to 3 cm 12/Pk Each for $28.00
Add to cart
Description
Description
  • For use in classroom identification labs and exercises
  • For use in lessons on mineral hardness, luster, and fracture
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Used in identification exercises, asking students to distinguish between it and other visually similar minerals such as feldspar or calcite
Specifications
Specifications
Milky Quartz
Mineral