Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Millennium Sciences™ CELL-VU™ CBC Disposable Hemacytometers

Ideal for manual erythrocyte, leukocyte, platelet, and eosinophil cell counts, the disposable CELL-VU Hemacytometer consists of a standard (3 × 1 in.) specially designed glass slide and coverslip

Manufacturer:  Millennium Sciences™ DRM 700

Catalog No. DRM700

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »


Description

Description

  • Disposing of the entire apparatus eliminates cleaning and minimizes clinician exposure to body fluids
  • Both the slide and coverslip are marked to ensure correct use
  • Each slide is dual-chambered (allowing for two determinations) whose printed inert surface creates a 0.18μL chamber volume
  • Centered on each coverslip is a laser-etched grid
  • The 3 × 3mm grid is a modified pattern of the Neubauer Ruling, divided into nine, 1 × 1mm counting areas
  • Each counting area is subdivided to facilitate specific, accurate results for the cell type being counted
  • The center square of the grid is subdivided into 100 counting areas and is used for erythrocyte and platelet counts

Order Info

Chamber depth is 20μm, optimal for cells to form in a monolayer. Each box includes 25 slides and 50 coverslips

Specifications

Specifications

Hemacytometer
0.50μL
50 Pk.
0.0007 in.
Ideal for manual erythrocyte, leukocyte, platelet, eosinophil and sperm cell counts
Laser-etched Centre
Yes
2.0
20μm
25 Slides and 50 coverslips
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit