Learn More
Honeywell™ Miller™ Black Rhino™ Self-Retracting Lifelines
Small, light 9 ft. (2.7m) cable lifeline
Manufacturer:  Honeywell™ CFL4Z7/9FT
Catalog No. 19-169-942
Add to cart
Description
Description
Ideal for concrete construction, welding, steel erection, food processing, and warehouse applications.
  • Stainless-steel wire rope endures daily rigors of harsh environments
  • Quick-activating, high-strength stainless-steel braking system—arrests free falls, providing a quick, safe rescue
  • Integral load indicator lets user know when unit must be removed from service and returned for repair
  • Corrosion-resistant stainless steel and aluminum components
  • Integral internal shock absorber consistently reduces fall forces to 900 lb. (408kg) or less for greater safety
  • Stainless-steel swivel and steel twist-lock carabiner
Compliance
Meets all applicable ANSI™, CSA, and OSHA standards, including ANSI Z359-2007.
Specifications
Specifications
900 lbf.
Stainless Steel, Aluminum
ANSI, CSA, OSHA, ANSI Z359-2007
Self retracting lifeline
Stainless Steel swivel and twist lock carabiner