Learn More
Honeywell™ Miller™ Turbo T-BAK™ Personal Fall Limiter
All-in-one, self-retracting lifeline/anchorage connector design reduces equipment required on the job site
Manufacturer:  Honeywell™ MFLT3/7.5FT
Catalog No. 19-175-510
Add to cart
Description
Description
Designed for anchoring below a user's harness back D-ring for greater safety
  • All-in-one, self-retracting lifeline/anchorage connector design reduces equipment required on the job site
  • Integrated shock absorber especially designed for tie-back applications with load indicator device
  • Cost-effective alternative to tie-back shock-absorbing lanyards
  • User-Rated to 400 lb. (181kg) capacity
  • 7.5 ft. (2.3m) working capacity
  • High-impact nylon housing
Certifications
Meets all applicable OSHA, ANSI, CSA and CE Marked standards, including the ANSI Z359.1 requirement that states only 24-inches (.62m) or less of lifeline extends out of the housing when the unit is fully retracted
Specifications
Specifications
7.5 ft.
Meets all applicable OSHA, ANSI, CSA and CE Marked standards
181 kg
2.3 m
400 lb.