Learn More
Honeywell Miller™ H500 Safety Harness Series Construction Standard
Synthesizes safety, quality and comfort. The harness is designed around ergonomic safety, usability, style and performance, aiming to keep workers safe, supported and make their jobs easier.
Supplier:  Honeywell H5CS221022
 View more versions of this product
Catalog No. 19-080-023
Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area.
Description
Description
  • Specially formulated webbing flexes to accomodate movement and body contours for optimum comfort around legs and shoulders
  • Highly breathable, lightweight, ergonomic shoulder/back padding minimizes overhead muscular fatigue
    • Keeps workers drier and cooler
  • Robust and ergonomic pressure-relief waist pad with adequate foam density for optimum lumbar support
  • Smart anti-tangle for hassle-free donning experience
  • User friendly resettable lanyard keepers
  • Clear identifiable smart impact indicators for easier inspection
  • Rated for up to a 420 lb. weight capacity
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Universal
Quick-connect leg buckles
Full body harness
Back and side
Anchorage connector, Self-retracting lifeline, Lanyard
Quick-connect chest buckles
OSHA 1926.502, ANSI Z359.11, CSA Z259.10
6.43 lb.
2.92 kg
Nylon, Steel
Product Suggestions
Product Suggestions