Honeywell Miller™ HP™ Fall Arrest and Retrieval Harnesses
Webbing coated with PTFE to extend service life
Manufacturer: Honeywell 850T58/XXLGK
 View more versions of this product
Catalog No. 19-120-2750
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Certifications/Compliance Meets all applicable OSHA, ANSI, and CSA requirements.
    Color Bright green
    Material Nylon Webbing; PTFE coating
    Size 2X-Large
    Buckle Style Qwik-Fit
    D-Ring Location Back; Side
    Superior water and oil repellency and protection against dirt, grime, spills and stains.
    • With lanyard ring, sub-pelvic strap, back strap and back or side D-rings
    • Bright green for high visibility and ease of inspection
    • Friction buckles on the shoulder straps
    (This style is also available with front D-ring, side D-rings and mating buckles on the shoulder straps. See below.) Models 850T and 850T-7 have mating buckles on both the leg and chest straps. Models 850T-4 and 850T-58 have tongue buckles on the leg straps and mating buckles on the chest strap.
  • Description & Specifications
Related Products
Product Content Feedback