Miller™ Non-Stretch Harnesses with Tongue-Buckle Leg Straps

Manufacturer: Honeywell Safety Products 80954/SYK

 View more versions of this product

Catalog No. 19-069-993A

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Product Type Subpelvic non-stretch full-body harness
    Style Subpelvic full body
    Buckle Style Leg Strap: Tongue; Shoulder Strap; Friction; Chest Strap: Mating
    Webbing Material 1.75 in. Polyester
    Capacity (English) 400 lb.
    Certifications/Compliance ANSI A10.32, ANSI Z359.1, OSHA 1926.502
    Color Black/Yellow
    For Use With (Application) Ideal for being suspended, raised or lowered, such as in tanks, manholes and pipe
    Size Small

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit