Learn More
Honeywell™ Miller™ Perfect Fit CRT Gloves
Perfect Fit CRT (Cut-Resistant Technology), provide ANSI cut level 4 cut resistance in even the lightest weight, CRT13 glove. CRT is a proprietary blend of cut-resistant fibers. Seamless knit gloves are soft and comfortable to wear, and offer excellent dexterity and abrasion resistance.
Manufacturer:  Honeywell™ CRTD13L
 View more versions of this product
Catalog No. 19-825-215
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Honeywell introduces its newest addition to the Perfect Fit, cut-resistant line. Perfect Fit CRT (Cut-Resistant Technology) offers excellent cut protection in a light weight seamless knit glove. Perfect Fit CRT's proprietary blend of cut resistant fibers offers 2 to 3 times the cut protection of other similar weight aramid gloves.
Compliance
Meets ANSI A4 (cut level) standards.
Specifications
Specifications
Level 4
agriculture, manufacturing, municipal services, Steel and Metals, transportation
With dot pattern palm coating
Black, Yellow
Knit
Women's
Light weight