Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Miller™ Perfect Fit CRT Gloves

Click to view available options
Size:
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Perfect Fit CRT (Cut-Resistant Technology), provide ANSI cut level 4 cut resistance. CRT is a proprietary blend of comfortable cut-resistant fibers featuring dexterity and abrasion resistance.

Perfect Fit CRT (Cut-Resistant Technology) offers cut protection in a light weight seamless knit glove. Perfect Fit CRT's proprietary blend of cut resistant fibers offers 2 to 3 times the cut protection of other similar weight aramid gloves

Color Black, Yellow
Cut Resistance Rating ANSI A4
Product Type Cut Resistant Gloves
Size 2X-Large
For Use With (Application) Agriculture, manufacturing, municipal services, steel and metals, transportation
Glove Cuff Style Knit