Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Miller™ Perfect Fit CRT Gloves

Perfect Fit CRT (Cut-Resistant Technology), provide ANSI cut level 4 cut resistance in even the lightest weight, CRT13 glove. CRT is a proprietary blend of cut-resistant fibers. Seamless knit gloves are soft and comfortable to wear, and offer excellent dexterity and abrasion resistance.

$3.71 - $37.45

Save up to 64%

Specifications

Color Black, Yellow
Cut Resistance Rating Level 4
For Use With (Application) agriculture, manufacturing, municipal services, Steel and Metals, transportation
Glove Cuff Style Knit
Weight Light weight
View More Specs

Products 7
Catalog Number Mfr. No. Size Style Price Quantity & Availability  
Catalog Number Mfr. No. Size Style Price Quantity & Availability  
19-825-202 View Documents Honeywell™
CRT13J
2X-Large Without dot pattern palm coating Pair for $3.71

Save 64% Reg : $10.25

Add to cart
 
19-825-201 View Documents Honeywell™
CRT13
Men's Without dot pattern palm coating Pair for $8.10
Add to cart
 
19-825-200 View Documents Honeywell™
CRT13L
Women's With outdot pattern palm coating Pair for $7.30
Add to cart
 
19-825-217 View Documents Honeywell™
CRTD13J
2X-Large With dot pattern palm coating Dozen of 12 for $37.45
Case of 12 DZ for $449.29
Add to cart
 
19-825-216 View Documents Honeywell™
CRTD13
Men's With dot pattern palm coating This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-825-215 View Documents Honeywell™
CRTD13L
Women's With dot pattern palm coating This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-825-218 View Documents Honeywell™
CRTD13XJ
3X-Large With dot pattern palm coating This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Honeywell introduces its newest addition to the Perfect Fit, cut-resistant line. Perfect Fit CRT (Cut-Resistant Technology) offers excellent cut protection in a light weight seamless knit glove. Perfect Fit CRT's proprietary blend of cut resistant fibers offers 2 to 3 times the cut protection of other similar weight aramid gloves.

Compliance

Meets ANSI A4 (cut level) standards.

Specifications

Specifications

Black, Yellow
agriculture, manufacturing, municipal services, Steel and Metals, transportation
Light weight
Level 4
Knit
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit