Honeywell Miller™ Revolution™ DualTech™ Harnesses

Harnesses with DualTech webbing offering safety and functionality with features developed to meet key user needs

Manufacturer: Honeywell RDTFDTBBDP/UBK

 View more versions of this product

Catalog No. 19-165-292

  Check Availability
Add to Cart

    Specifications

    Style Full-body harness
    Buckle Style Tongue Buckle Legs
    D-Ring Location Front; Side
    Webbing Material DualTech™/polyester
    Includes Removable belt and pad, integrated accessory system
    Certifications/Compliance OSHA, ANSI and CSA
    Color Black
    For Use With (Application) Construction, manufacturing and transportation fall protection
    Size Universal

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit