Miller™ Revolution™ Harness Accessories

Attach easily to PivotLink™ connection on harness

Manufacturer: Honeywell Safety Products RIAT6/6

 View more versions of this product

Catalog No. 19-130-4113

  Check Availability
Add to Cart

    Specifications

    For Use With (Equipment) Miller Revolution Harnesses
    Product Type Two pocket nail, tool pouch
    Material Nylon
    Color Black
    Includes Water bottle holder, belt with pad and pivotlink attachment, five accessory connectors

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit