Honeywell Miller™ Revolution™ Harnesses

Design based on key user-identified needs

Manufacturer: Honeywell RDTTBDP/UBK

 View more versions of this product

Catalog No. 19-034-867

  Check Availability
Add to Cart

    Specifications

    Product Type Revolution Harness
    Style With Side D-rings and pad
    D-Ring Location Side
    Material DualTech Webbing
    Buckle Style Tongue-Buckle Leg Straps
    Color Black
    For Use With (Application) Construction, Fall Protection
    Size Universal

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit