Honeywell Miller™ Revolution™ Harnesses

Design based on key user-identified needs

Manufacturer: Honeywell R10CNTBBDP/UGN

 View more versions of this product

Catalog No. 17-987-603

  Check Availability
Add to Cart

    Specifications

    Product Type Revolution Harness
    Buckle Style Tongue-Buckle Leg Straps
    D-Ring Location Side
    For Use With (Application) Construction, Fall Protection
    Size Universal
    Style With Removable belt, Side D-Rings and Pad
    Color Black
    Material Type 10 Webbing

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit