Miller™ SofStop™ Shock-Absorbing Lanyard
Manufacturer: Honeywell Safety Products 928LS18INYL
 View more versions of this product
Catalog No. 17-987-499A
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Product Type Shock Absorbing Pack with Locking Snap and D Ring
    Arresting Force (English) 900 lb. max.
    Certifications/Compliance Meet OSHA 1926.502, 1910.66; ANSI A10.32 and CSA Z259.11
    Color Yellow and Black
  • Description & Specifications
Related Products
Product Content Feedback