Honeywell Miller™ StretchStop™ Lanyard with SofStop Shock Absorber
Combination of integral SofStop Shock Absorber with stretchable lanyard design
Manufacturer: Honeywell 8798RSSZ7/6FTGNC
 View more versions of this product
Catalog No. 19-034-894
  Check Availability
Add to Cart
    Specifications
    Product Type Web Lanyard, ANSI Z359 Compliant, Two legs
    Leg Configuration Double-Leg
    Color Mirrored
    Connector Types Locking Snap Hook and 2 Locking Rebar Hooks
    Length (English) 6 ft.
  • Description & Specifications
Related Products
Product Content Feedback