Honeywell Miller™ TranScendor™ Cable Climbing Systems
Permanently installed, flexible cable system provides continuous fall protection for climbing fixed ladders
Manufacturer: Honeywell TRS/40FT
 View more versions of this product
Catalog No. 19-074-541
  Check Availability
Add to Cart
    Specifications
    Length (English) 40 ft.
    Length (Metric) 1219cm
    Includes Manual pass-through
    Certifications/Compliance OSHA, ANSI, CSA
    For Use With (Application) Ideal for agriculture, construction, manufacturing, municipal services, Oil and gas, Steel and Metals, utilities
  • Description & Specifications
Related Products
Product Content Feedback