Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Miller™ TurboLite™ Personal Fall Limiters

Never need a shock-absorbing lanyard again with the 6-ft lightweight personal fall limiters rated for a 400-lb worker. Affordable alternative to shock-absorbing lanyards.

Manufacturer:  Honeywell™ MFL1Z7/9FT

 View more versions of this product

Catalog No. 19-066-226


Add to cart

Description

Description

  • Eliminates need for different fall protection equipment to address fall clearance
  • Extremely compact and lightweight
  • Provide 1.8m (6 ft.) or 2.7m (9 ft.) working capacity lifeline
  • Attach directly to harness back D-ring for greater mobility and versatility
  • Engineered webbing for greater abrasion resistance and long service life
  • High-strength, impact-resistant nylon housing for maximum durability
  • Built-in swivel prevents lifeline from twisting
  • Rated for up to 181.4kg (400 lb.)

No annual factory recertification is required unless unit is involved in a fall.

Compliance

Meets OSHA 1910.66 and OSHA 1926.502 (d); ANSI™ A10.32 and ANSI Z359.1-2007; CSA Z259.2.2.(Type I) and EN 360 requirements.

Specifications

Specifications

Safety
High impact Nylon
ANSI Z359-2007
181.43kg
2.7m
1.13kg
TurboLite personal fall limiter
Steel carabiner for unit, Steel snap hook for lanyard end, 3600 lb. gate
400 lb.
9 ft.
2.5 lb.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit