Learn More
MilliporeSigma™ Millex™-FH Syringe Filter Unit, Hydrophobic PTFE, 0.45 μm Edge Available on GSA/VA Contract for Federal Government customers only.
Edge
Protect vacuum sources and tank/bioreactor venting from contamination and moisture damage. Millex™ Filters: Inlet and Outlet for Latex Tubing are constructed of sturdy polypropylene with hydrophobic PTFE membranes.
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
SLFH05000
View Documents
MilliporeSigma™
SLFH05000
100Pack
Each for $1,209.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
SLFH05010 Edge Available on GSA/VA Contract for Federal Government customers only.
View Documents
MilliporeSigma™
SLFH05010
10Pack
Each for $184.10
N/A
Edge
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Protect vacuum sources and tank/bioreactor venting from contamination and moisture damage. Millex™ Filters: Inlet and Outlet for Latex Tubing are constructed of sturdy polypropylene with hydrophobic PTFE membranes.
Millex™ syringe filter units with hydrophobic Fluoropore™ or Millipore Express™ PLUS PES membrane are ideal for in-line sterilization of gases and venting sterile containers, and filters with Fluoropore™ membrane can also be used for sterilizing or clarifying organic solutions. There are also specialized filter units to protect hemodialysis transducers from blood and moisture. The 50 mm Millex™ filter units are especially useful for vacuum line protection. All units are bi-directional.
Features & Benefits:
  • Specialized filter units protect hemodialysis transducers from blood and moisture
  • 50 mm Millex™ units ideal for vacuum line protection
  • All Millex™ Hydrophobic PTFE units are bi-directional in flow
Applications:
Sterilizing Gases, Vacuum Line Protection, Venting Sterile Containers, Sterilizing and Clarifying Organic Solutions