Learn More
MilliporeSigma™ Millex™ Syringe Filter, MCE, 0.22 μm
For removing particulates, reducing signal-to-noise ratios and maintaining flat baselines
$867.50
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
SLGS02510
View Documents
MilliporeSigma™
SLGS02510
1000Pack
Pack of 1000 for $3,340.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
SLGS025NB
View Documents
MilliporeSigma™
SLGS025NB
250Pack
Each for $867.50
Add to cart
Description
Description
For removing particulates, reducing signal-to-noise ratios and maintaining flat baselines
Non-Sterile Millex™ Syringe Filters with MCE Membrane:
  • Housing is made from a low-extractable polyvinylidene chloride (PVC).
Applications:
Clarification of Aqueous Solutions, General Laboratory Filtration