Learn More
MilliporeSigma™ Millex™ Syringe Filter, Nylon, Automation Compatible, 0.2 μm
Trouble-free performance on a variety of automated systems
$2530.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
SLGNDZ5NK
View Documents
MilliporeSigma™
SLGNDZ5NK
1000Pack
Each for $2,530.00
Add to cart
SLGNDZ5NZ
View Documents
MilliporeSigma™
SLGNDZ5NZ
200Pack
Pack of 200 for $726.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Trouble-free performance on a variety of automated systems
Automation-Compatible, Non-Sterile Millex™ Syringe Filters:
  • Designed to provide efficient filtration for dissolution and other automated sample preparation applications
  • Domed housing ensures reliable delivery of filters
  • Available in a variety of membrane types and pore sizes, including units with prefilters for particulate-laden samples.
  • Available in either bulk or delivery tubes for use with automated filter-changing systems
  • Luer-Lok™ connection optimized for precise alignment and fit
Applications:
  • Drug Dissolution Testin
  • Automated Sample Preparation
  • Clarification of Solutions Containing High Particle Levels