Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Millex™ Syringe Filter, PES, 0.22 μm Available on GSA/VA Contract for Federal Government customers only.

For removing particulates, reducing signal-to-noise ratios and maintaining flat baselines

$321.00 - $2460.00

Specifications

Pore Size 0.22 μm
Sterility Non-sterile
For Use With (Application) Analytical sample prep
Housing Material PP
View More Specs

Products 4
Catalog Number Mfr. No. Diameter (Metric) Filter Quantity Material Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Filter Quantity Material Volume (Metric) Price Quantity & Availability  
SLGPX13NK Available on GSA/VA Contract for Federal Government customers only.
View Documents
MilliporeSigma™
SLGPX13NK
13 mm 1000Pack Hydrophilic PES <10 mL
Pack of 1000 for $2,060.00
Add to cart
 
SLGPX13NL Available on GSA/VA Contract for Federal Government customers only.
View Documents
MilliporeSigma™
SLGPX13NL
13 mm 100Pack Hydrophilic PES <10 mL
Pack of 100 for $321.00
Add to cart
 
SLGP033NB Available on GSA/VA Contract for Federal Government customers only.
View Documents
MilliporeSigma™
SLGP033NB
33 mm 250Pack Millipore Express PLUS (PES) <200 mL
Pack of 250 for $951.00
Add to cart
 
SLGP033NK Available on GSA/VA Contract for Federal Government customers only.
View Documents
MilliporeSigma™
SLGP033NK
33 mm 1000Pack Millipore Express PLUS (PES) <200 mL
Pack of 1000 for $2,460.00
Add to cart
 
Description

Description

For removing particulates, reducing signal-to-noise ratios and maintaining flat baselines

Overview of Non-Sterile Millex™ Syringe Filters with PES Membrane:
  • Non-sterile Millex™ syringe filters with Millipore Express™ PLUS polyethersulfone (PES) membrane provide fast filtration of aqueous and mild organic solutions.
  • Low protein binding to minimize interaction with your sample and maximize recovery
  • Polypropylene housing, offering a low level of extractables and broad chemical compatibility

Applications:
Buffer Filtration, Clarification of Aqueous Solutions

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

0.22 μm
Non-sterile
Analytical sample prep
PP
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit