Learn More
MilliporeSigma™ Milli-Q™ 7010-7015 Purification Cartridge Kit
Catalog No. ix70xxpkt1h Shop All MilliporeSigma Products
Click to view available options
Type:
IX 7010-7015 Purification Cartridge Kit
IX H 7010-7015 Purification Cartridge Kit
For optimal protection of the Reverse Osmosis membrane for use with Milli-Q™ IX 7010 and Milli-Q™ IX 7015 systems, and advanced protection against airborne contaminants for Milli-Q™ IQ tank top.
Water Purification Consumables Kit, compatible with the Milli-Q™ IX 7010 and Milli-Q™ IX 7010 systems and includes:
  • IPAK GARD™ 10-15 Pretreatment Pack
  • Activated Carbon for protection against chlorine, organics, bacteria
  • Depth Filter for protection against particulates and colloids
  • Tank Vent Filter
  • Activated Carbon for volatile organics removal
  • Soda Lime for CO2 removal
  • Membrane Filter to stop particulates and bacteria
TRUSTED_SUSTAINABILITY
Specifications
Capacity 25 L, 50 L and/or 100 L
Flow Rate 3 L/hr. (Milli-Q™ IX 7003), 5 L/hr. (Milli-Q™ IX 7005), 10 L/hr. (Milli-Q™ IX 7010), 15 L/hr. (Milli-Q™ IX 7015)
For Use With (Equipment) Milli-Q™ IX 7003/05/10/15 Water Purification System
Type IX H 7010-7015 Purification Cartridge Kit
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.