Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ MILLICUP™-FLEX Disposable Filtration Kit
MILLICUP™-FLEX Disposable Filtration Kit combines the convenience of disposable filtration with the flexibility and compatibility of traditional vacuum filtration glassware.
$61.25 - $122.00
Specifications
Autoclavable | Not Autoclavable |
---|---|
Color | Clear with blue adapter |
Material | Polypropylene |
Sterility | Non-sterile |
Outlet Fitting | Universal Bottletop |
Description
- Compatible with organic and aqueous solvents
- Ergonomic, clampless design
- Reduce contamination risk
- Filter directly into vacuum-rated storage bottles
- Easy access to membrane after filtration (membranes sold separately)
- Fully recyclable components
- Compatible with 47 mm membranes
- GL45 threads
- Solvent-resistant
Specifications
Not Autoclavable | |
Polypropylene | |
Universal Bottletop | |
Bottle Top Filter |
Clear with blue adapter | |
Non-sterile | |
Universal Bottletop |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title