Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Millipak™ Sterile Filters for Milli-Q™ IQ 7000 Ultrapure Water System
Purification cartridges
$356.00 - $360.00
Specifications
For Use With (Equipment) | Milli-Q IQ 7000 Ultrapure Water System |
---|---|
Type | Sterile Filter |
Purity Meter Included | No |
Description
Our Lab Water purification system portfolio offers a broad range of pure and ultrapure water purification systems and services designed for scientists working in pharmaceutical, clinical, academic, industrial, research, and government laboratories — in both validated and non-validated environments.
- 0.22μm membrane filter for particulate-free and bacteria-free water at the point of dispense for the Milli-Q IQ 7000 system
- Sterile unit
- Delivered hermetically sealed.
Specifications
Milli-Q IQ 7000 Ultrapure Water System | |
Sterile Filter | |
No |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title