Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mineral oil, for spectroscopy, suitable for nujol mull preparations for IR-spect., ACROS Organics™

Click to view available options
Quantity:
6mL
1L
2.5L
10L
Packaging:
Glass Bottle
Plastic drum
8042-47-5
C16H10N2Na2O7S2
452.363
MFCD00131611
AEOVEGJBKQQFOP-DDVLFWKVSA-L
acid orange 10,wool orange 2g,orange g,c.i. acid orange 10,c.i. orange g,c.i. food orange 4,light orange g,colacid orange g,dolkwal orange g,hexacol orange g
9566064
disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate
C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
8042-47-5
C16H10N2Na2O7S2
452.363
MFCD00131611
AEOVEGJBKQQFOP-DDVLFWKVSA-L
acid orange 10,wool orange 2g,orange g,c.i. acid orange 10,c.i. orange g,c.i. food orange 4,light orange g,colacid orange g,dolkwal orange g,hexacol orange g
9566064
disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate
C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
For IR spectroscopy
Flash Point 185°C
Infrared Spectrum Authentic
Color Colorless
Quantity 10L
Density 0.8510g/mL
Viscosity 14-19 mm2/s (40°C)
Specific Gravity 0.851
Packaging Plastic drum
Refractive Index 1.4620 to 1.4730
Merck Index 14, 7186
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in benzene, chloroform, ether,, carbon dislulfide, petroleum ether, oils, miscible with wax and fats, insoluble in alcohol
Physical Form Oily Liquid
Grade Spectroscopy
Chemical Name or Material Mineral oil
Show More Show Less

GHS P Statement May be fatal if swallowed and enters airways.

GHS P Statement IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Do NOT induce vomiting.

Danger

EINECSNumber : 232-455-8

RTECSNumber : PY8047000

TSCA : TSCA