Learn More
Scientific IndustriesSupplier Diversity Partner MINI 300 ORB-GENIE FLASK 120V DFS Item
Manufacturer:  Scientific IndustriesSupplier Diversity Partner SIM1600
120V. Heavy duty, low profile, compact footprint Orbital Shaker for Standard Flask Clamps with variable shaking speed up to 300RPM. Ideal for applications using various size flasks or beakers. Clamps sold separately.
Catalog No. 50-112-6491