Learn More
Estes Industries, LLC Mini Altitrak
SDPSCI_ED
Take learning to new heights with this low-cost technology solution for tracking model rocket altitude. These small, easy to transport altitude tracking devices allow for students to track flight data, analyze and graph. Coming in a 3 pack.
Manufacturer:  Estes Industries, LLC 002226
Catalog No. S68263
Add to cart
Description
Description
  • The Mini AltiTrak uses a gravity protractor to present the angle from your body to the apogee of your rocket flight.
  • With the distance from the launchpad to your body, use simple geometry to calculate the height of your rocket flight, then challenge your students to engineer solutions to optimize their rocket’s altitude
Specifications
Specifications
Blue
Rocket Altitude
10.5 in.
14.25 in.
Altitude Tracker
1 in.
6 in.
Model Rockets
MINI ALTITRAK™ ALTITUDE TRACKER
Plastic
2.9 oz.