Justrite™ Mini-Berm Flex Spill Control Tray

Compact, portable spill control for harsh and hazardous chemicals

Manufacturer: Justrite™ 28442

 View more versions of this product

Catalog No. 19-111-028

  Check Availability
Add to Cart

    Specifications

    Capacity (English) 30 gal.
    Capacity (Metric) 113.5L
    Dimensions (L x W x H) 1.8 x 1.5 x 0.15m
    Height (Metric) 1.5m
    Includes 1 in. NPT drain fitting/utility strap
    Length (Metric) 0.9m
    Material Modified PVC coated fabric
    Width (Metric) 0.9m
    Product Type Spill Control Tray

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit