Learn More
Thermo Scientific™ Mini Capillary Unions
Thermo Scientific™ Mini Capillary Unions are for use with capillary GC columns.
$127.20
Specifications
Type Capillary Union
View More Specs
Products 2
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
03-050-244
View Documents
Thermo Scientific™
290GU498
0.22 to 0.32 mm ID Capillary GC Column
Each for $127.20
Only null left
Add to Cart
03-050-245
View Documents
Thermo Scientific™
290GU499
0.53 mm ID Capillary GC Column
Pack of 10 for $127.20
Only null left
Add to Cart
Description
Description
Thermo Scientific™ Mini Capillary Unions are for use with capillary GC columns.
Kit contains one union and five ferrules
Includes:
Union, 1; Ferrules, 5
Specifications
Specifications
Capillary Union
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.